Sodium borohydride 4.4 M in 14 M sodium hydroxide solution
Supplier: BeanTown Chemical
Synonyms:
Sodium tetrahydridoborate, Sodium tetrahydroborate
139595-100ML
139595-2L
139595-500ML
BT139595-100MLEA
47.27
USD
BT139595-100ML
BT139595-2L
BT139595-500ML
Sodium borohydride 4.4 M in 14 M sodium hydroxide solution
Sodium borohydride
CAS: 16940-66-2; EC No: 241-004-4; MDL No: MFCD00003518
UN No: UN3320; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: NaBH4; MW: 37.83
Boiling Point: 132.2°
Density (g/mL): 1.375
Air Sensitive
- Air Sensitive
- Liquid
Formula:
NaBH₄ MW: 37.83 g/mol Boiling Pt: 132.2 °C Melting Pt: 400 Density: 1.375 g/mL Flash Pt: 69 |
MDL Number:
MFCD00003518 CAS Number: 16940-66-2 EINECS: 241-004-4 UN: 3320 ADR: 4.3 II Merck Index: 13,08664 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...