Arsenic(III) oxide ≥99.99%
Supplier: BeanTown Chemical
Synonyms:
di-Arsenic trioxide
139570-10G
139570-250G
139570-50G
BT139570-10GEA
59.5
USD
BT139570-10G
BT139570-250G
BT139570-50G
Arsenic(III) oxide ≥99.99%
Arsenic (III) oxide
CAS: 1327-53-3; EC No: 215-481-4; MDL No: MFCD00003433; RTECS: CG3325000
UN No: UN1561; Haz Class: 6.1; Packing Group: II
-100 Mesh Powder; Molecular Formula: As2O3; MW: 197.84
Boiling Point: 465° (sublimes)
- RTECS: CG3325000
- -100 Mesh Powder
Formula:
As₂O₃ Boiling Pt: 465 °C (subl.) |
MDL Number:
MFCD00003433 CAS Number: 1327-53-3 EINECS: 215-481-4 UN: 1561 ADR: 6.1,II |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...