Tetrabutylammonium iodide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Tetra-n-butylammonium iodide, TBAI
139565-1KG
139565-250G
139565-50G
BT139565-1KGEA
344
USD
BT139565-1KG
BT139565-250G
BT139565-50G
Tetrabutylammonium iodide ≥98%
Tetrabutylammonium iodide
CAS: 311-28-4; EC No: 206-220-5; MDL No: MFCD00011636; RTECS: BS5450000
Crystalline/Powder; Linear Formula: [CH3(CH2)3]4NI; Molecular Formula: C16H36IN; MW: 369.38
Melting Point: 141-143°
Hygroscopic , Light Sensitive
- RTECS: BS5450000
- Hygroscopic, Light Sensitive
- Crystalline/Powder
Formula:
C₁₆H₃₆IN Melting Pt: 141…143 °C |
MDL Number:
MFCD00011636 CAS Number: 311-28-4 EINECS: 206-220-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...