Dicarbonylcyclopentadienylcobalt

Supplier: BeanTown Chemical

Synonyms: Cyclopentadienylcobalt dicarbonyl, Dicarbonylcyclopentadienylcobalt(I)

139560-1G 139560-5G
BT139560-1GEA 45.63 USD
BT139560-1G BT139560-5G
Dicarbonylcyclopentadienylcobalt
Dicarbonylcyclopentadienylcobalt

CAS: 12078-25-0; EC No: 235-139-8; MDL No: MFCD00009589 UN No: UN3281; Haz Class: 6.1; Packing Group: II Liquid; Molecular Formula: C7H5CoO2 ; MW: 180.05 Boiling Point: 139-140°/710 mmHg; Flash point: 27°C (81°F) Air Sensitive


  • Air Sensitive
  • Liquid

Formula: C₇H₅CoO₂
MW: 180.05 g/mol
Boiling Pt: 139…140 °C/710 mmHg
Flash Pt: 27 °C (81 °F)
MDL Number: MFCD00009589
CAS Number: 12078-25-0
EINECS: 235-139-8
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR