Dicarbonylcyclopentadienylcobalt
Supplier: BeanTown Chemical
Synonyms:
Cyclopentadienylcobalt dicarbonyl, Dicarbonylcyclopentadienylcobalt(I)
139560-1G
139560-5G
BT139560-1GEA
45.63
USD
BT139560-1G
BT139560-5G
Dicarbonylcyclopentadienylcobalt
Dicarbonylcyclopentadienylcobalt
CAS: 12078-25-0; EC No: 235-139-8; MDL No: MFCD00009589
UN No: UN3281; Haz Class: 6.1; Packing Group: II
Liquid; Molecular Formula: C7H5CoO2 ; MW: 180.05
Boiling Point: 139-140°/710 mmHg; Flash point: 27°C (81°F)
Air Sensitive
- Air Sensitive
- Liquid
Formula:
C₇H₅CoO₂ Boiling Pt: 139…140 °C/710 mmHg Flash Pt: 27 °C (81 °F) |
MDL Number:
MFCD00009589 CAS Number: 12078-25-0 EINECS: 235-139-8 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...