Cobalt (III) acetylacetonate
Supplier: BeanTown Chemical
Synonyms:
Cobalt (III) tris(2,4-pentanedionato-O,O'), Cobalt (III) 2,4-pentanedionate, Cobalt tris(acetylacetonate), Tris(2,4-pentanedionato-O,O')cobalt (III)
139540-100G
139540-25G
139540-5G
BT139540-100GEA
134.63
USD
BT139540-100G
BT139540-25G
BT139540-5G
Cobalt (III) acetylacetonate
Cobalt (III) acetylacetonate
CAS: 21679-46-9; EC No: 244-527-6; MDL No: MFCD00013488
Powder; Molecular Formula: C15H21CoO6 ; MW: 356.26
Melting Point: 210-213°
Hygroscopic, Moisture Sensitive
- Hygroscopic, Moisture Sensitive
- Powder
Formula:
C₁₅H₂₁CoO₆ Melting Pt: 210…213 °C |
MDL Number:
MFCD00013488 CAS Number: 21679-46-9 EINECS: 244-527-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...