Benzo[b]thiophene-3-carboxylic acid ≥96%
Supplier: BeanTown Chemical
Synonyms:
1-Benzothiophene-3-carboxylic acid
139520-1G
139520-25G
139520-5G
BT139520-1GEA
97
USD
BT139520-1G
BT139520-25G
BT139520-5G
Benzo[b]thiophene-3-carboxylic acid ≥96%
Benzo[b]thiophene-3-carboxylic acid
CAS: 5381-25-9; MDL No: MFCD01846406
Solid; Molecular Formula: C9H6O2S; MW: 178.22
Melting Point: 174-177°
- Solid
Formula:
C₉H₆O₂S Melting Pt: 174…177 °C |
MDL Number:
MFCD01846406 CAS Number: 5381-25-9 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...