Diethyl (hydroxymethyl)phosphonate ≥85%, Technical Grade

Supplier: BeanTown Chemical

139420-25G 139420-5G
BT139420-25GEA 108.57 USD
BT139420-25G BT139420-5G
Diethyl (hydroxymethyl)phosphonate ≥85%, Technical Grade
Diethyl (hydroxymethyl)phosphonate

CAS: 3084-40-0; EC No: 221-391-6; MDL No: MFCD00014418 Liquid; Molecular Formula: C5H13O4P; MW: 168.13 Boiling Point: 124-126°/3 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.14; Refractive Index: 1.444


  • Refractive index: 1.444
  • Liquid

Formula: C₅H₁₃O₄P
Boiling Pt: 124…126 °C/3 mmHg
Density: 1.14 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00014418
CAS Number: 3084-40-0
EINECS: 221-391-6

Order Now

Specification Test Results

Assay 85%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR