Dextrin

Supplier: BeanTown Chemical

Synonyms: Dextrin

139135-100G 139135-500G
BT139135-100GEA 42.57 USD
BT139135-100G BT139135-500G
Dextrin
Dextrin

CAS: 9004-53-9; EC No: 232-675-4; MDL No: MFCD00081554; RTECS: HH9450000 Powder Hygroscopic


  • RTECS: HH9450000
  • Hygroscopic
  • Powder

Formula: (C₆H₁₂O₆)n
MDL Number: MFCD00081554
CAS Number: 9004-53-9
EINECS: 232-675-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR