Copper (I) sulfide ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Dicopper sulfide, Dicopper sulphide, Chacolcite, Copper(i)sulfide, Copper(i)sulphide

139120-1KG 139120-250G 139120-50G
BT139120-1KGEA 879 USD
BT139120-1KG BT139120-250G BT139120-50G
Copper (I) sulfide ≥99.5% (trace metals basis)
Copper (I) sulphide

CAS: 22205-45-4; EC No: 244-842-9; MDL No: MFCD00016062 -200 Mesh Powder; Molecular Formula: Cu2S; MW: 159.15 Melting Point: 1100° Density (g/mL): 5.6


  • -200 Mesh Powder

Formula: Cu₂S
Melting Pt: 1100 °C
Density: 5.6 g/mL
MDL Number: MFCD00016062
CAS Number: 22205-45-4
EINECS: 244-842-9

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR