Gold ≥99.95% (trace metals basis), spherical powder APS 5.5-9.0 µm
Supplier: BeanTown Chemical
139090-1G
139090-5G
BT139090-1GEA
224
USD
BT139090-1G
BT139090-5G
Gold ≥99.95% (trace metals basis), spherical powder APS 5.5-9.0 µm
Gold
CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436; RTECS: MD5420000
Powder, S.A. 0.05-0.20 m2/g ; Molecular Formula: Au; MW: 196.97
Melting Point: 1063°; Boiling Point: 2808°
Density (g/mL): 19.3
- RTECS: MD5420000
- Powder, S.A. 0.05-0.20 m²/g
Formula:
Au Boiling Pt: 2808 °C Melting Pt: 1063 °C Density: 19.3 g/mL |
MDL Number:
MFCD00003436 CAS Number: 7440-57-5 EINECS: 231-165-9 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...