Bismuth ≥99.5% (trace metals basis), powder -100 mesh
Supplier: BeanTown Chemical
139040-100G
139040-2KG
139040-500G
BT139040-100GEA
45.98
USD
BT139040-100G
BT139040-2KG
BT139040-500G
Bismuth ≥99.5% (trace metals basis), powder -100 mesh
Bismuth
CAS: 7440-69-9; EC No: 231-177-4; MDL No: MFCD00134033; RTECS: EB2600000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder, apparent density 3.8-4.6g/cm3; Molecular Formula: B; MW: 208.98
Melting Point: 271°; Boiling Point: 1560°
Density (g/mL): 9.8
- RTECS: EB2600000
- Powder, apparent density 3.8-4.6g/cm3
Formula:
Bi Boiling Pt: 1560 °C Melting Pt: 271 °C Density: 9.8 g/mL |
MDL Number:
MFCD00134033 CAS Number: 7440-69-9 EINECS: 231-177-4 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...