Chlorotriethylsilane ≥98%

Supplier: BeanTown Chemical

Synonyms: Triethylchlorosilane

138990-10G 138990-250G 138990-50G
BT138990-10GEA 43.33 USD
BT138990-10G BT138990-250G BT138990-50G
Chlorotriethylsilane ≥98%
Chlorotriethylsilane

CAS: 994-30-9; EC No: 213-615-6; MDL No: MFCD00000507 UN No: UN2986; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: (CH3CH2)3SiCl; Molecular Formula: C6H15ClSi; MW: 150.73 Boiling Point: 142-144°; Flash point: 30°C (86°F) Density (g/mL): 0.898; Refractive Index: 1.43 Moisture Sensitive


  • Refractive index: 1.43
  • Moisture Sensitive
  • Liquid

Formula: C₆H₁₅ClSi
Boiling Pt: 142…144 °C
Density: 0.898 g/mL
Flash Pt: 30 °C (86 °F)
MDL Number: MFCD00000507
CAS Number: 994-30-9
EINECS: 213-615-6
UN: 2986
ADR: 8,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR