Silver cyanide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Argentous cyanide
138940-100G
138940-25G
BT138940-100GEA
630
USD
BT138940-100G
BT138940-25G
Silver cyanide ≥99%
Silver cyanide
CAS: 506-64-9; EC No: 208-048-6; MDL No: MFCD00003409; RTECS: VW3850000
UN No: UN1684; Haz Class: 6.1; Packing Group: II
Powder; Linear Formula: AgCN; MW: 133.89
Melting Point: 320° (decomposes)
Density (g/mL): 3.95; Refractive Index: 1.685
Light Sensitive
- RTECS: VW3850000
- Refractive index: 1.685
- Light Sensitive
- Powder
Formula:
AgCN Melting Pt: 320 °C (dec.) Density: 3.95 g/mL |
MDL Number:
MFCD00003409 CAS Number: 506-64-9 EINECS: 208-048-6 UN: 1684 ADR: 6.1,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...