5-Nonanol ≥98%
Supplier: BeanTown Chemical
138860-25G
BT138860-25GEA
44
USD
BT138860-25G
5-Nonanol ≥98%
5-Nonanol
CAS: 623-93-8; EC No: 210-820-2; MDL No: MFCD00021944
Liquid; Molecular Formula: C9H20O; MW: 144.25
Boiling Point: 195°; Flash point: 77°C (171°F)
Density (g/mL): 0.821; Refractive Index: 1.429
- Refractive index: 1.429
- Liquid
Formula:
C₉H₂₀O Boiling Pt: 195 °C Density: 0.821 g/mL Flash Pt: 77 °C (171 °F) |
MDL Number:
MFCD00021944 CAS Number: 623-93-8 EINECS: 210-820-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...