Lead(II) perchlorate trihydrate ≥97.0% ACS
Supplier: BeanTown Chemical
138855-100G
138855-500G
BT138855-100GEA
48
USD
BT138855-100G
BT138855-500G
Lead(II) perchlorate trihydrate ≥97.0% ACS
Lead (II) perchlorate trihydrate
CAS: 13453-62-8; EC No: 237-125-7; MDL No: MFCD00149817
UN No: UN1470; Haz Class: 5.1 (6.1); Packing Group: II
Crystalline; Linear Formula: Pb(ClO4)2·3H2O; MW: 460.15
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Cl₂O₈Pb·3H₂O |
MDL Number:
MFCD00149817 CAS Number: 13453-62-8 EINECS: 237-125-7 UN: 1470 ADR: 5.1,II |
Specification Test Results
Assay | 97.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...