Titanium ≥99.99% (trace metals basis), foil, Thickness 0.127 mm
Supplier: BeanTown Chemical
138745-100X100
138745-25X25MM
138745-50X50MM
BT138745-100X1EA
466.43
USD
BT138745-100X1
BT138745-25X25
BT138745-50X50
Titanium ≥99.99% (trace metals basis), foil, Thickness 0.127 mm
Titanium
CAS: 7440-32-6; EC No: 231-142-3; MDL No: MFCD00011264; RTECS: XR1700000
Solid; Molecular Formula: Ti; MW: 47.87
Melting Point: 1660°; Boiling Point: 3287°
Density (g/mL): 4.5
- RTECS: XR1700000
- Solid
Formula:
Ti Boiling Pt: 3287 °C Melting Pt: 1660 °C Density: 4.5 g/mL |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...