Sulfuric acid 0.2 N in water volumetric solution

Supplier: BeanTown Chemical

138725-1L 138725-20L 138725-4L
BT138725-1LEA 42.72 USD
BT138725-1L BT138725-20L BT138725-4L
Sulfuric acid 0.2 N in water volumetric solution
Sulfuric acid

CAS: 7664-93-9; EC No: 231-639-5; MDL No: MFCD00064589; RTECS: WS5600000 UN No: UN2796; Haz Class: 8; Packing Group: II Liquid; Linear Formula: H2SO4; MW: 98.08


  • RTECS: WS5600000
  • Liquid

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR