Sulfuric acid 93-98%, Environmental Grade
Supplier: BeanTown Chemical
138715-2.5L
138715-500ML
BT138715-2.5LEA
187
USD
BT138715-2.5L
BT138715-500ML
Sulfuric acid 93-98%, Environmental Grade
Sulfuric acid
CAS: 7664-93-9; EC No: 231-639-5; MDL No: MFCD00064589; RTECS: WS5600000
UN No: UN1830; Haz Class: 8; Packing Group: II
Liquid, single sub-boiling quartz distillation; Linear Formula: H2SO4; MW: 98.08
Melting Point: 3°; Boiling Point: 290°
Density (g/mL): 1.84
- RTECS: WS5600000
- Liquid, single sub-boiling quartz distillation
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.84 g/mL |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8 II |
Specification Test Results
Assay | 93-98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...