Chromium(VI) oxide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Chromic anhydride, Chromium trioxide

138590-100G 138590-25G 138590-500G
BT138590-100GEA 165.84 USD
BT138590-100G BT138590-25G BT138590-500G
Chromium(VI) oxide ≥99.9%
Chromium (VI) oxide

CAS: 1333-82-0; EC No: 215-607-8; MDL No: MFCD00010952; RTECS: GB6650000 UN No: UN1463; Haz Class: 5.1 (6.1,8); Packing Group: II Crystalline; Molecular Formula: CrO3; MW: 99.99 Melting Point: 196° (decomposes) Density (g/mL): 2.7 Hygroscopic


  • RTECS: GB6650000
  • Hygroscopic
  • Crystalline

Formula: CrO₃
MW: 99.99 g/mol
Melting Pt: 195 (dec.)
Density: 2.7 g/mL
MDL Number: MFCD00010952
CAS Number: 1333-82-0
EINECS: 215-607-8
UN: 1463
ADR: 5.1 II
Merck Index: 13,02256

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR