EDTA (ethylenediamine tetraacetic acid) ≥99.4% ACS
Supplier: BeanTown Chemical
138580-100G
138580-2.5KG
138580-500G
BT138580-100GEA
34
USD
BT138580-100G
BT138580-2.5KG
BT138580-500G
EDTA (ethylenediamine tetraacetic acid) ≥99.4% ACS
EDTA (Ethylenediamine tetraacetic acid)
CAS: 60-00-4; EC No: 200-449-4; MDL No: MFCD00003541; RTECS: AH4025000
Powder; Molecular Formula: C10H16N2O8; MW: 292.24
Melting Point: 250° (decomposes)
- RTECS: AH4025000
- Powder
Formula:
C₁₀H₁₆N₂O₈ Melting Pt: 250 °C (dec.) |
MDL Number:
MFCD00003541 CAS Number: 60-00-4 EINECS: 200-449-4 |
|
Specification Test Results
Assay | 99.4% min. |
Residue on ignition | ≤0.20% |
Calcium (Ca) | ≤0.001% |
Magnesium (Mg) | ≤5ppm |
Heavy metals (as Pb) | ≤0.001% |
Iron (Fe) | ≤0.005% |
Nitrilotriacetic acid | ≤0.1% |
Insoluble in dilute ammonium hydroxide | ≤0.005% |
Appearance | White/off-white powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...