EDTA (ethylenediamine tetraacetic acid) ≥99.4% ACS

Supplier: BeanTown Chemical

138580-100G 138580-2.5KG 138580-500G
BT138580-100GEA 34 USD
BT138580-100G BT138580-2.5KG BT138580-500G
EDTA (ethylenediamine tetraacetic acid) ≥99.4% ACS
EDTA (Ethylenediamine tetraacetic acid)

CAS: 60-00-4; EC No: 200-449-4; MDL No: MFCD00003541; RTECS: AH4025000 Powder; Molecular Formula: C10H16N2O8; MW: 292.24 Melting Point: 250° (decomposes)


  • RTECS: AH4025000
  • Powder

Formula: C₁₀H₁₆N₂O₈
Melting Pt: 250 °C (dec.)
MDL Number: MFCD00003541
CAS Number: 60-00-4
EINECS: 200-449-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.4% min.
Residue on ignition ≤0.20%
Calcium (Ca) ≤0.001%
Magnesium (Mg) ≤5ppm
Heavy metals (as Pb) ≤0.001%
Iron (Fe) ≤0.005%
Nitrilotriacetic acid ≤0.1%
Insoluble in dilute ammonium hydroxide ≤0.005%
Appearance White/off-white powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR