2-Pentanone ≥99%
Supplier: BeanTown Chemical
Synonyms:
Methyl Propyl Ketone
138500-100G
138500-25G
138500-500G
BT138500-100GEA
29.94
USD
BT138500-100G
BT138500-25G
BT138500-500G
2-Pentanone ≥99%
2-Pentanone
CAS: 107-87-9; EC No: 203-528-1; MDL No: MFCD00009400; RTECS: SA7875000
UN No: UN1249; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3COCH2CH2CH3; Molecular Formula: C5H10O; MW: 86.13
Melting Point: -78°; Boiling Point: 101-105°; Flash point: 7°C (45°F)
Density (g/mL): 0.809; Refractive Index: 1.390
- RTECS: SA7875000
- Refractive index: 1.390
- Liquid
Formula:
C₅H₁₀O Boiling Pt: 101…105 °C Melting Pt: –78 °C Density: 0.809 g/mL Flash Pt: 7 °C (45 °F) |
MDL Number:
MFCD00009400 CAS Number: 107-87-9 EINECS: 203-528-1 UN: 1249 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...