Catechol violet
Supplier: BeanTown Chemical
138490-25G
138490-5G
BT138490-25GEA
155
USD
BT138490-25G
BT138490-5G
Catechol violet
Catechol violet
CAS: 115-41-3; EC No: 204-088-3; MDL No: MFCD00005868
Powder; Molecular Formula: C19H14O7S; MW: 386.38
- Powder
Formula:
C₁₉H₁₄O₇S |
MDL Number:
MFCD00005868 CAS Number: 115-41-3 EINECS: 204-088-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...