Sodium iodide ≥99%
Supplier: BeanTown Chemical
Synonyms:
Hydriodic acid sodium salt, Sodium monoiodide, Ioduril
138360-100G
138360-2.5KG
138360-500G
BT138360-100GEA
51.38
USD
BT138360-100G
BT138360-2.5KG
BT138360-500G
Sodium iodide ≥99%
Sodium iodide
CAS: 7681-82-5; EC No: 231-679-3; MDL No: MFCD00003532; RTECS: WB6475000
Crystalline; Linear Formula: NaI; MW: 149.89
Melting Point: 661°; Boiling Point: 1300-1304°
Density (g/mL): 3.67; Refractive Index: 1.775
Air Sensitive, Hygroscopic, Light Sensitive
- RTECS: WB6475000
- Refractive index: 1.775
- Air Sensitive, Hygroscopic, Light Sensitive
- Crystalline
Formula:
NaI MW: 149.89 g/mol Boiling Pt: 1300 Melting Pt: 662 Density: 3.67 g/mL |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 EINECS: 231-679-3 UN: 3077 ADR: 9 III Merck Index: 13,08705 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...