Sodium iodide ≥99%

Supplier: BeanTown Chemical

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

138360-100G 138360-2.5KG 138360-500G
BT138360-100GEA 51.38 USD
BT138360-100G BT138360-2.5KG BT138360-500G
Sodium iodide ≥99%
Sodium iodide

CAS: 7681-82-5; EC No: 231-679-3; MDL No: MFCD00003532; RTECS: WB6475000 Crystalline; Linear Formula: NaI; MW: 149.89 Melting Point: 661°; Boiling Point: 1300-1304° Density (g/mL): 3.67; Refractive Index: 1.775 Air Sensitive, Hygroscopic, Light Sensitive


  • RTECS: WB6475000
  • Refractive index: 1.775
  • Air Sensitive, Hygroscopic, Light Sensitive
  • Crystalline

Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300
Melting Pt: 662
Density: 3.67 g/mL
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9 III
Merck Index: 13,08705

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR