Iodine(V) oxide ≥98%

Supplier: BeanTown Chemical

Synonyms: di-Iodine pentaoxide, Iodopentoxide, di-Iodine pentoxide

138355-100G 138355-25G
BT138355-100GEA 273 USD
BT138355-100G BT138355-25G
Iodine(V) oxide ≥98%
Iodine (V) oxide

CAS: 12029-98-0; EC No: 234-740-2; MDL No: MFCD00011356 UN No: UN3085; Haz Class: 5.1 (8); Packing Group: II Crystalline; Molecular Formula: I2O5; MW: 333.81 Density (g/mL): 5.08 Hygroscopic , Light Sensitive


  • Hygroscopic, Light Sensitive
  • Crystalline

Formula: I₂O₅
MW: 333.81 g/mol
Density: 5.08 g/mL
MDL Number: MFCD00011356
CAS Number: 12029-98-0
EINECS: 234-740-2
UN: 3085
ADR: 5.1 II
Merck Index: 12,05040

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR