Strontium carbonate ≥99.99%
Supplier: BeanTown Chemical
138340-100G
138340-25G
138340-500G
BT138340-100GEA
279
USD
BT138340-100G
BT138340-25G
BT138340-500G
Strontium carbonate ≥99.99%
Strontium carbonate
CAS: 1633-05-2; EC No: 216-643-7; MDL No: MFCD00011250
Powder; Linear Formula: SrCO3; MW: 147.63
Melting Point: 1100° (decomposes)
Density (g/mL): 3.7
- Powder
Formula:
CO₃Sr Melting Pt: 1100 °C (dec.) Density: 3.7 g/mL |
MDL Number:
MFCD00011250 CAS Number: 1633-05-2 EINECS: 216-643-7 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...