Benzophenone ≥99%
Supplier: BeanTown Chemical
Synonyms:
Diphenylmethanone, Diphenyl ketone
138280-1KG
138280-250G
138280-5KG
BT138280-1KGEA
68.76
USD
BT138280-1KG
BT138280-250G
BT138280-5KG
Benzophenone ≥99%
Benzophenone
CAS: 119-61-9; EC No: 204-337-6; MDL No: MFCD00003076; RTECS: DI9950000
UN No: UN3077; Haz Class: 9; Packing Group: III
Flakes; Molecular Formula: C13H10O; MW: 182.22
Melting Point: 47-51°; Boiling Point: 305°
- RTECS: DI9950000
- Flakes
Formula:
C₁₃H₁₀O Boiling Pt: 305 °C Melting Pt: 47…51 °C |
MDL Number:
MFCD00003076 CAS Number: 119-61-9 EINECS: 204-337-6 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...