Sodium perchlorate 98.0-102.0% ACS
Supplier: BeanTown Chemical
138240-100G
138240-500G
BT138240-100GEA
46.2
USD
BT138240-100G
BT138240-500G
Sodium perchlorate 98.0-102.0% ACS
Sodium perchlorate
CAS: 7601-89-0; EC No: 231-511-9; MDL No: MFCD00003480
UN No: UN1502; Haz Class: 5.1; Packing Group: II
Crystalline/Flakes; Linear Formula: NaClO4; MW: 122.46
Melting Point: 468°
Density (g/mL): 2.499
Hygroscopic
- Hygroscopic
- Crystalline/Flakes
Formula:
ClNaO₄ Melting Pt: 468 °C Density: 2.499 g/mL |
MDL Number:
MFCD00003480 CAS Number: 7601-89-0 EINECS: 231-511-9 UN: 1502 ADR: 5.1,II |
Specification Test Results
Assay | 98.0-102.0% |
pH of 5% solution at 25°C | 6.0-8.0 |
Insoluble matter | ≤0.005% |
Chloride (Cl) | ≤0.003% |
Sulfate (SO4) | ≤0.002% |
Calcium (Ca) | ≤0.02% |
Iron (Fe) | ≤0.0005% |
Heavy metals (as Pb) | ≤0.0005% |
Potassium (K) | ≤0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...