Pyrite (FeS₂), grains, Diameter 1.5-4.8 mm

Supplier: BeanTown Chemical

138185-1G 138185-25G 138185-5G
BT138185-1GEA 25 USD
BT138185-1G BT138185-25G BT138185-5G
Pyrite (FeS₂), grains, Diameter 1.5-4.8 mm
Pyrite (FeS₂)

CAS: 1309-36-0; EC No: 215-167-7; MDL No: MFCD00064690 Solid; Molecular Formula: FeS2; MW: 119.98 Density (g/mL): 5.02; Refractive Index: 3.08


  • Refractive index: 3.08
  • Solid

Formula: FeS₂
Density: 5.02 g/mL
MDL Number: MFCD00064690
CAS Number: 1309-36-0
EINECS: 215-167-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR