Phenylphosphonic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
Phenylphosphonic Acid
138150-100G
138150-500G
BT138150-100GEA
58
USD
BT138150-100G
BT138150-500G
Phenylphosphonic acid ≥98%
Phenylphosphonic acid
CAS: 1571-33-1; EC No: 212-220-6; MDL No: MFCD00002136; RTECS: TD4393000
UN No: UN3261; Haz Class: 8; Packing Group: III
Crystalline; Molecular Formula: C6H7O3P; MW: 158.09
Melting Point: 162-164°
- RTECS: TD4393000
- Crystalline
Formula:
C₆H₇O₃P Melting Pt: 162…164 °C |
MDL Number:
MFCD00002136 CAS Number: 1571-33-1 EINECS: 212-220-6 UN: 3261 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...