Graphite ≥99.9%, felt, Thickness 1.12 cm (0.441 in)

Supplier: BeanTown Chemical

138105-10X10CM 138105-25X25CM
BT138105-10X10EA 46 USD
BT138105-10X10 BT138105-25X25
Graphite ≥99.9%, felt, Thickness 1.12 cm (0.441 in)
Graphite

CAS: 7782-42-5; EC No: 231-955-3; MDL No: MFCD00144065 Solid; Molecular Formula: C; MW: 12.011


  • Solid

Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR