Platinum, Plasma Standard Solution, Pt 1000 µg/ml
Supplier: BeanTown Chemical
138080-500ML
BT138080-100MLEA
205.61
USD
BT138080-100ML
BT138080-500ML
BT138080-50ML
Platinum, Plasma Standard Solution, Pt 1000 µg/ml
Standards
Cationic Standards
Platinum Standards
EC No: 231-595-7
Haz class: 8
Packing group: II
GHS symbol: GHS05
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...