5-Bromo-2-hydroxypyrimidine ≥98%
Supplier: BeanTown Chemical
Synonyms:
5-Bromopyrimidin-2(1H)-one, 5-Bromo-2-pyrimidinone
138050-1G
138050-5G
BT138050-1GEA
71
USD
BT138050-1G
BT138050-5G
5-Bromo-2-hydroxypyrimidine ≥98%
5-Bromo-2-hydroxypyrimidine
CAS: 38353-06-9; EC No: 253-896-2; MDL No: MFCD00233952
Solid; Molecular Formula: C4H3BrN2O; MW: 174.98
Melting Point: 230° (decomposes)
- Solid
Formula:
C₄H₃BrN₂O Melting Pt: 230 °C (dec.) |
MDL Number:
MFCD01318956 CAS Number: 38353-06-9 EINECS: 253-896-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...