Ytterbium ≥99.9% (trace rare earth metals basis), foil, Thickness 1.0 mm
Supplier: BeanTown Chemical
138020-25X25MM
138020-50X50MM
BT138020-25X25EA
180
USD
BT138020-25X25
BT138020-50X50
Ytterbium ≥99.9% (trace rare earth metals basis), foil, Thickness 1.0 mm
Ytterbium
CAS: 7440-64-4; EC No: 231-173-2; MDL No: MFCD00011286
Solid; Molecular Formula: Yb; MW: 173.04
Melting Point: 824°; Boiling Point: 1196°
Density (g/mL): 6.54
- Solid
Formula:
Yb Boiling Pt: 1196 °C Melting Pt: 824 °C Density: 6.54 g/mL |
MDL Number:
MFCD00011286 CAS Number: 7440-64-4 EINECS: 231-173-2 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...