Tungsten, gauze woven from wire 100 mesh Ø 0.025 mm
Supplier: BeanTown Chemical
138005-150X150
138005-300X300
138005-75X75MM
BT138005-150X1EA
522
USD
BT138005-150X1
BT138005-300X3
BT138005-75X75
Tungsten, gauze woven from wire 100 mesh Ø 0.025 mm
Tungsten
CAS: 7440-33-7; EC No: 231-143-9; MDL No: MFCD00011461
Wire Cloth; Molecular Formula: W; MW: 183.84
Melting Point: 3410°; Boiling Point: 5660°
Density (g/mL): 19.3
- Wire Cloth
Formula:
W Boiling Pt: 5660 °C Melting Pt: 3410 °C Density: 19.3 g/mL |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...