Copper(II) nitrate hemipentahydrate 98.0-102.0% ACS

Supplier: BeanTown Chemical

Synonyms: Cupric Nitrate Hydrate, Copper dinitrate hemipentahydrate, Cupric nitrate hemi(pentahydrate), Cupric nitrate 2.5-hydrate, Cupric nitrate hemipentahydrate

137915-100G 137915-2KG 137915-500G
BT137915-100GEA 39.16 USD
BT137915-100G BT137915-2KG BT137915-500G
Copper(II) nitrate hemipentahydrate 98.0-102.0% ACS
Copper (II) nitrate hemipentahydrate

CAS: 19004-19-4; EC No: 221-838-5; MDL No: MFCD00149671; RTECS: GL7875000 UN No: UN3085; Haz Class: 5.1 (8); Packing Group: III Crystalline/Lump; Linear Formula: Cu(NO3)2·2.5H2O; MW: 232.59 Density (g/mL): 2.32 Hygroscopic


  • RTECS: GL7875000
  • Hygroscopic
  • Crystalline/Lump

Formula: Cu(NO₃)₂·2.5H₂O
Density: 2.32 g/mL
MDL Number: MFCD02253001
CAS Number: 19004-19-4
EINECS: 221-838-5
UN: 3085
ADR: 5.1,III

Order Now

Specification Test Results

Assay 98.0-102.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR