Diethyl (bromodifluoromethyl)phosphonate ≥97%

Supplier: BeanTown Chemical

137900-1G 137900-25G 137900-5G
BT137900-1GEA 26 USD
BT137900-1G BT137900-25G BT137900-5G
Diethyl (bromodifluoromethyl)phosphonate ≥97%
Diethyl (bromodifluoromethyl)phosphonate

CAS: 65094-22-6; MDL No: MFCD00069118 Liquid; Molecular Formula: C5H10BrF2O3P; MW: 267.01 Boiling Point: 40-41°/0.05 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.503; Refractive Index: 1.417


  • Refractive index: 1.417
  • Liquid

Formula: C₅H₁₀BrF₂O₃P
MW: 267.01 g/mol
Boiling Pt: 40…41 °C/0.05 mmHg
Density: 1.503 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00069118
CAS Number: 65094-22-6

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR