Tin(IV) oxide ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

137885-100G 137885-2KG 137885-500G
BT137885-100GEA 40 USD
BT137885-100G BT137885-2KG BT137885-500G
Tin(IV) oxide ≥99.9%
Tin (IV) oxide

CAS: 18282-10-5; EC No: 242-159-0; MDL No: MFCD00011244; RTECS: XQ4000000 10 Micron Powder; Linear Formula: SnO2; MW: 150.71 Melting Point: 1630°; Boiling Point: 1800-1900° Density (g/mL): 6.95; Refractive Index: 1.997


  • RTECS: XQ4000000
  • Refractive index: 1.997
  • <10 Micron Powder

Formula: O₂Sn
Boiling Pt: 1800…1900 °C
Melting Pt: 1630 °C
Density: 6.95 g/mL
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR