Perfluoro-tert-butanol ≥99%

Supplier: BeanTown Chemical

Synonyms: Perfluoro-t-butanol, Nonafluoro-tert-butyl alcohol, 1,1,1,3,3,3-Hexafluoro-2-trifluoromethyl-2-propanol, Perfluoro-tert-butyl alcohol

137865-1G 137865-25G 137865-5G
BT137865-1GEA 91 USD
BT137865-1G BT137865-25G BT137865-5G
Perfluoro-tert-butanol ≥99%
Perfluoro-tert-butanol

CAS: 2378-02-1; EC No: 219-157-3; MDL No: MFCD00042092; RTECS: UB6452700 UN No: UN2810; Haz Class: 6.1; Packing Group: II Liquid; Linear Formula: (CF3)3COH; Molecular Formula: C4HF9O; MW: 236.04 Melting Point: -17°; Boiling Point: 45° Density (g/mL): 1.693; Refractive Index: 1.300


  • RTECS: UB6452700
  • Refractive index: 1.300
  • Liquid

Formula: C₄HF₉O
MW: 236.04 g/mol
Boiling Pt: 84
Melting Pt: -17
Density: 1.693 g/mL
MDL Number: MFCD00042092
CAS Number: 2378-02-1
EINECS: 219-157-3
UN: 2810
ADR: 6.1 II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR