Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥99%
Supplier: BeanTown Chemical
Synonyms:
2-Hydrazinyl-N,N,N-trimethyl-2-oxoethanaminium chloride
137860-100G
BT137860-100GEA
55.65
USD
BT137860-100G
Girard's reagent T (carbazoylmethyltrimethylammonium chloride) ≥99%
Girard's reagent T (Carbazoylmethyltrimethylammonium chloride)
CAS: 123-46-6; EC No: 204-629-3; MDL No: MFCD00012009; RTECS: CP3150000
Crystalline; Linear Formula: [(CH3)3NCH2CONHNH2]Cl; Molecular Formula: C5H14ClN3O; MW: 167.64
Melting Point: 188-192° (decomposes)
Hygroscopic
- RTECS: CP3150000
- Hygroscopic
- Crystalline
Formula:
C₅H₁₄ClN₃O MW: 167.64 g/mol Melting Pt: 188…192 °C (dec.) |
MDL Number:
MFCD00012009 CAS Number: 123-46-6 EINECS: 204-629-3 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...