Rhodium(II) acetate dimer ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
137830-1G
137830-250MG
BT137830-1GEA
605.55
USD
BT137830-1G
BT137830-250MG
Rhodium(II) acetate dimer ≥99.99% (trace metals basis)
Rhodium (II) acetate dimer
CAS: 15956-28-2; EC No: 240-084-8; MDL No: MFCD00003538; RTECS: VI9361000
Powder; Linear Formula: Rh2(OOCCH3)4; MW: 441.99
Melting Point: 205°
- RTECS: VI9361000
- Powder
Formula:
C₈H₁₂O₈Rh₂ Melting Pt: 205 °C |
MDL Number:
MFCD00003538 CAS Number: 15956-28-2 EINECS: 240-084-8 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...