3-Bromo-5-methylpyridine ≥97%
Supplier: BeanTown Chemical
137820-1G
137820-5G
BT137820-1GEA
138
USD
BT137820-1G
BT137820-5G
3-Bromo-5-methylpyridine ≥97%
3-Bromo-5-methylpyridine
CAS: 3430-16-8; MDL No: MFCD01646141
Liquid; Molecular Formula: C6H6BrN ; MW: 172.02
Flash point: 90°C (194°F)
Density (g/mL): 1.4869; Refractive Index: 1.562
- Refractive index: 1.562
- Liquid
Formula:
C₆H₆BrN Density: 1.4869 g/mL Flash Pt: 90 °C (194 °F) |
MDL Number:
MFCD01646141 CAS Number: 3430-16-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...