Zinc(II) nitrate hexahydrate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Zinc nitrate hexahydrate
137785-10KG
137785-2KG
137785-500G
BT137785-10KGEA
420
USD
BT137785-10KG
BT137785-2KG
BT137785-500G
Zinc(II) nitrate hexahydrate ≥99%
Zinc (II) nitrate hexahydrate
CAS: 10196-18-6; EC No: 231-943-8; MDL No: MFCD00149889; RTECS: ZH4775000
UN No: UN1514; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: Zn(NO3)2·6H2O; MW: 297.49
Melting Point: 36°
Density (g/mL): 2.065
- RTECS: ZH4775000
- Crystalline
Formula:
ZnN₂O₆·6H₂O MW: 297.49 g/mol Melting Pt: 36 Density: 2.065 g/mL |
MDL Number:
MFCD00149889 CAS Number: 10196-18-6 EINECS: 231-943-8 UN: 1514 ADR: 5.1 II Merck Index: 12,10273 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...