Sodium selenite, anhydrous ≥99%
Supplier: BeanTown Chemical
137760-100G
137760-500G
BT137760-100GEA
80.23
USD
BT137760-100G
BT137760-500G
Sodium selenite, anhydrous ≥99%
Sodium selenite
CAS: 10102-18-8; EC No: 233-267-9; MDL No: MFCD00003489; RTECS: VS7350000
UN No: UN2630; Haz Class: 6.1; Packing Group: I
Crystalline/Powder; Linear Formula: Na2SeO3; MW: 172.94
Melting Point:
- RTECS: VS7350000
- Hygroscopic
- Crystalline/Powder
Formula:
Na₂SeO₃ Melting Pt: >350 °C |
MDL Number:
MFCD00003489 CAS Number: 10102-18-8 EINECS: 233-267-9 UN: 2630 ADR: 6.1,I |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...