1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Cyclen tetrahydrochloride
137700-1G
137700-250MG
137700-5G
BT137700-1GEA
217
USD
BT137700-1G
BT137700-250MG
BT137700-5G
1,4,7,10-Tetraazacyclododecane tetrahydrochloride ≥98%
1,4,7,10-Tetraazacyclododecane tetrahydrochloride
CAS: 10045-25-7; MDL No: MFCD00068484
UN No: UN3077; Haz Class: 9; Packing Group: III
Solid; Molecular Formula: C8H20N4·4HCl; MW: 318.11
Melting Point: 293° (decomposes)
Hygroscopic
- Hygroscopic
- Solid
Formula:
C₈H₂₄Cl₄N₄ MW: 318.12 g/mol Melting Pt: 293 °C (dec.) |
MDL Number:
MFCD00068484 CAS Number: 10045-25-7 ADR: 9 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...