Iron(III) chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Iron trichloride, Ferric chloride
137640-100G
137640-1KG
BT137640-100GEA
39.92
USD
BT137640-100G
BT137640-1KG
Iron(III) chloride ≥98%
Iron (III) chloride
CAS: 7705-08-0; EC No: 231-729-4; MDL No: MFCD00011005; RTECS: LJ9100000
UN No: UN1773; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: FeCl3; MW: 162.21
Melting Point: 304°
Hygroscopic
- RTECS: LJ9100000
- Hygroscopic
- Crystalline
Formula:
FeCl₃ Melting Pt: 304 °C |
MDL Number:
MFCD00011005 CAS Number: 7705-08-0 EINECS: 231-729-4 UN: 1773 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...