Iron(III) chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Iron trichloride, Ferric chloride

137640-100G 137640-1KG
BT137640-100GEA 39.92 USD
BT137640-100G BT137640-1KG
Iron(III) chloride ≥98%
Iron (III) chloride

CAS: 7705-08-0; EC No: 231-729-4; MDL No: MFCD00011005; RTECS: LJ9100000 UN No: UN1773; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: FeCl3; MW: 162.21 Melting Point: 304° Hygroscopic


  • RTECS: LJ9100000
  • Hygroscopic
  • Crystalline

Formula: FeCl₃
MW: 162.21 g/mol
Boiling Pt: 315
Melting Pt: 260
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 1773
ADR: 8 III
Merck Index: 13,04048

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR