Potassium nitrate ≥99.99% (trace metals basis)

Supplier: BeanTown Chemical

137625-100G 137625-25G
BT137625-100GEA 218 USD
BT137625-100G BT137625-25G
Potassium nitrate ≥99.99% (trace metals basis)
Potassium nitrate

CAS: 7757-79-1; EC No: 231-818-8; MDL No: MFCD00011409; RTECS: TT3700000 UN No: UN1486; Haz Class: 5.1; Packing Group: III Crystalline; Molecular Formula: KNO3; MW: 101.11 Melting Point: 334°; Boiling Point: 400° (decomposes) Density (g/mL): 2.109 Hygroscopic


  • RTECS: TT3700000
  • Hygroscopic
  • Crystalline

Formula: KNO₃
Boiling Pt: 400 °C (dec.)
Melting Pt: 334 °C
Density: 2.109 g/mL
MDL Number: MFCD00011409
CAS Number: 7757-79-1
EINECS: 231-818-8
UN: 1486
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR